Description
Thermo Scientific™ Remel RPR Card Test Kit is a nontreponemal flocculation test for detection of reagin in human serum when used in conjunction with a treponemal test for presumptive diagnosis of syphilis.
Specifications
Specifications
| Description | RPR Card Test Kit/500Pk. |
| Quantity | 500 Tests/Kit |
| Target Organism Class | Treponema pallidum |