Learn More
Thermo Scientific™ SABHI™ Agar w/Chloramphenicol, Cycloheximide
GSA_VA
Catalog No. R01754 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
20mm x 113mm Tube
Monoplate Deep Pour
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
R01754 Monoplate Deep Pour
R08697 20mm x 113mm Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R01754 Supplier Thermo Scientific™ Supplier No. R01754
Add to Cart
Add to Cart
This item is not returnable. View return policy
Selectively cultivate and isolate pathogenic fungi using SABHI™ Agar w/Chloramphenicol, Cycloheximide.
Selectively cultivate and isolate pathogenic fungi with Thermo Scientific™ Remel™ SABHI™ Agar w/Chloramphenicol, Cycloheximide. In 1967, Gorman developed SABHI Agar by combining Sabouraud Dextrose Agar and Brain Heart Infusion Agar. This combined formulation increased recovery of pathogenic fungi1. Selectivity can be achieved by the addition of different antibiotics. Sabouraud Dextrose Agar helps in the cultivation of dermatophytes and Brain Heart Infusion Agar helps to isolate fungi from clinical specimens2.
SABHI Agar w/Chloramphenicol, Cycloheximide is a solid medium recommended for use in qualitative procedures for cultivation and isolation of pathogenic fungi.
This medium is made selective by adding chloramphenicol and cycloheximide that inhibit a range of gram-positive and gram-negative bacteria and saprophytic fungi respectively.
  • Ready-to-use – Convenience of prepared media
  • Available in two formats: as a deep pour monoplate and a 20 x 133 mm tube
  • Selective – Chloramphenicol is an broad spectrum antibiotic which inhibits a wide range of gram-positive and gram-negative bacteria. Cycloheximide inhibits rapidly growing saprophytic fungi that may overgrow slower-growing organisms as well as growth of other pathogens such as Cryptococcus neoformans, some Candida species, some Aspergillus species and Zygomycetes
  • Conforms to CLSI Standards3
SABHI Agar w/Chloramphenicol, Cycloheximide contains dextrose, casein peptone, meat peptone, brain heart infusion solids, sodium chloride, disodium phosphate, agar, demineralized water, chloramphenicol and cycloheximide. Dextrose serves as the carbohydrate source. Sheep blood provide essential growth factors for fastidious fungi. Chloramphenicol and gentamicin inhibit gram-positive and gram-negative bacteria.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description SABHI Agar w/Chloramphenicol, Cycloheximide (Sabouraud Dextrose w/Brain Heart Infusion Agar)
Format Monoplate Deep Pour
Product Type Agar Plate
Quantity 10/Pk.