Learn More
Description
Cultivate and isolate pathogenic fungi using SABHI™ (Sabouraud Dextrose with Brain Heart Infusion Agar) (5% sheep blood).
Cultivate and isolate pathogenic fungi with Thermo Scientific™ Remel™ SABHI™ (Sabouraud Dextrose with Brain Heart Infusion Agar) (5% sheep blood). In 1967, Gorman developed SABHI Agar by combining Sabouraud Dextrose Agar and Brain Heart Infusion Agar. This combined formulation increased recovery of pathogenic fungi1. Selectivity can be achieved by the addition of different antibiotics. Sabouraud Dextrose Agar helps in the cultivation of dermatophytes and Brain Heart Infusion Agar helps to isolate fungi from clinical specimens2.
SABHI (Sabouraud Dextrose with Brain Heart Infusion Agar) (5% sheep blood) is a solid medium that is recommended for use in qualitative procedures for cultivation and isolation of pathogenic fungi.
- Ready-to-use – Convenience of prepared media
- Available in in two formats – as a deep pour monoplate and a 20 x 8.5 mL slant.
- Sheep blood provides essential growth factors that increase the recovery of fastidious fungi
- Conforms to CLSI Standards3
SABHI (Sabouraud Dextrose with Brain Heart Infusion Agar) (5% sheep blood) contains dextrose, casein peptone, meat peptone, brain heart infusion solids, sodium chloride, disodium phosphate, agar, demineralized water and sheep blood. Dextrose serves as the carbohydrate source. Sheep blood provide essential growth factors for fastidious fungi.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Description | SABHI Agar w/Blood (Sabouraud Dextrose w/Brain Heart Infusion Agar) (5% sheep blood) |
| Format | Monoplate Deep Pour |
| Product Type | Agar |
| Packaging Type | 10/Pk. |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.