Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SABHI™ Agar w/Sheep Blood

Catalog No. R08696
Change view
Click to view available options
Quantity:
10/Pk.
20 x 8.5 mL Slant
Format:
20mm x 113mm Tube
Monoplate Deep Pour
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R08696 20 x 8.5 mL Slant 20mm x 113mm Tube
R01752 10/Pk. Monoplate Deep Pour
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R08696 Supplier Thermo Scientific™ Supplier No. R08696
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cultivate and isolate pathogenic fungi using SABHI™ (Sabouraud Dextrose with Brain Heart Infusion Agar) (5% sheep blood).

Cultivate and isolate pathogenic fungi with Thermo Scientific™ Remel™ SABHI™ (Sabouraud Dextrose with Brain Heart Infusion Agar) (5% sheep blood). In 1967, Gorman developed SABHI Agar by combining Sabouraud Dextrose Agar and Brain Heart Infusion Agar. This combined formulation increased recovery of pathogenic fungi1. Selectivity can be achieved by the addition of different antibiotics. Sabouraud Dextrose Agar helps in the cultivation of dermatophytes and Brain Heart Infusion Agar helps to isolate fungi from clinical specimens2.

SABHI (Sabouraud Dextrose with Brain Heart Infusion Agar) (5% sheep blood) is a solid medium that is recommended for use in qualitative procedures for cultivation and isolation of pathogenic fungi.

  • Ready-to-use – Conve
TRUSTED_SUSTAINABILITY

Specifications

Description SABHI Agar w/Blood (Sabouraud Dextrose w/Brain Heart Infusion Agar) (5% sheep blood)
Format 20mm x 113mm Tube
Product Type Agar
Packaging Type 20/Pk.
Quantity 20 x 8.5 mL Slant
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.