Learn More
Thermo Scientific™ SABHI™ Agar (Sabouraud Dextrose with Brain Heart Infusion Agar)
GSA_VA
Catalog No. R01750
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
20/Pk.
Format:
20mm x 113mm Tube
Monoplate Deep Pour
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R01750 10/Pk. Monoplate Deep Pour
R01749 100/Pk. Monoplate Deep Pour
R08690 100/Pk. 20mm x 113mm Tube
R08692 20/Pk. 20mm x 113mm Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. R01750 Supplier Thermo Scientific™ Supplier No. R01750
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Cultivate and isolate pathogenic fungi with this selective, ready-to-use medium.
Cultivate and isolate pathogenic fungi with Thermo Scientific™ Remel™ SABHI™ Agar. Sabouraud Dextrose Agar was developed by Sabouraud for cultivation of dermatophytes1. Brain Heart Infusion Agar is useful for primary isolation of fungi from clinical specimens2. In 1967, Gorman developed SABHI Agar combining Sabouraud Dextrose Agar and Brain Heart Infusion Agar. This combined formulation was found to increase the recovery of pathogenic fungi.
Pathogenic fungi have an enormous impact on human health. Most people are aware of some of the superficial infections caused by fungi. These include skin and nail infections such as athlete's foot and ringworm, predominantly caused by a dermatophyte. More seriously, certain fungal species can cause invasive diseases in humans.
Use SABHI™ Agar for the cultivation and isolation of pathogenic fungi.
  • The combined formulation of Brain Heart Infusion Agar and Sabouraud Dextrose Agar gives improved recovery of pathogenic fungi.
  • Ready-to-use convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description SABHI Agar
Format Monoplate Deep Pour
Product Type Agar Plate
Quantity 10/Pk.