Learn More
Thermo Scientific™ Remel™ Sabouraud Dextrose Agar, Emmons w/Chloramphenicol Bactiflask™
Catalog No. R10272 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20/Pk.
Format:
1 oz. TCF
Catalog No. Quantity Format
R10272 20/Pk. 1 oz. TCF
1 options
This item is not returnable. View return policy
Catalog No. R10272 Supplier Thermo Scientific™ Supplier No. R10272
This item is not returnable. View return policy
Thermo Scientific™ Remel Sabouraud Dextrose Agar, Emmons w/Chloramphenicol Bactiflask is used for selective isolation of yeast and fungi.
Thermo Scientific™ Remel Sabouraud Dextrose Agar, Emmons w/Chloramphenicol Bactiflask is used for selective isolation of yeast and fungi.
Chloramphenicol inhibits some gram-positive and gram-negative bacteria.
TRUSTED_SUSTAINABILITY
Specifications
Description Sabouraud Dextrose Agar, Emmons w/Chloramphenicol Bactiflask™
Format 1 oz. TCF
Product Type Agar
Quantity 20/Pk.