Learn More
Thermo Scientific™ Sabouraud Dextrose Agar, Emmons with Chloramphenicol and Cycloheximide
Shop All Thermo Scientific Products
Description
Isolate and cultivate pathogenic yeast and molds from clinical specimens with this selective, ready-to-use medium.
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Remel™ Sabouraud Dextrose Agar, Emmons with Chloramphenicol and Cycloheximide. Sabouraud Dextrose Agar, Emmons is a medium with an increased pH of 7.0 and reduced concentration of dextrose from 40 to 20 g/l2, compared to the original composition described in 1892 by Sabouraud1-2.
- Chloramphenicol inhibits some gram-positive and gram-negative bacteria
- Cycloheximide inhibits rapid growing saprophytic fungi
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Sabouraud Dextrose Agar, Emmons w/Chloramphenicol, Cycloheximide |
| Format | Monoplate Deep Pour |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |