Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Sabouraud Dextrose Agar, Emmons with Chloramphenicol and Cycloheximide
Click to view available options
Format:
20mm x 113mm Tube
Monoplate Deep Pour
Description
Isolate and cultivate pathogenic yeast and molds from clinical specimens with this selective, ready-to-use medium.
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Remel™ Sabouraud Dextrose Agar, Emmons with Chloramphenicol and Cycloheximide. Sabouraud Dextrose Agar, Emmons is a medium with an increased pH of 7.0 and reduced concentration of dextrose from 40 to 20 g/l2, compared to the original composition described in 1892 by Sabouraud1-2.
- Chloramphenicol inhibits some gram-positive and gram-negative bacteria
- Cycloheximide inhibits rapid growing saprophytic fungi
Specifications
Specifications
| Description | Sabouraud Dextrose Agar, Emmons w/Chloramphenicol, Cycloheximide |
| Format | Monoplate Deep Pour |
| Product Type | Agar |
| Quantity | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction