Learn More
Thermo Scientific™ Sabouraud Dextrose Agar, Emmons w/Chloramphenicol
GSA_VA
Catalog No. R01770 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
20/Pk.
Format:
F - slant
Monoplate Deep Pour
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R01770 10/Pk. Monoplate Deep Pour
R08724 100/Pk. F - slant
R08726 20/Pk. F - slant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R01770 Supplier Thermo Scientific™ Supplier No. R01770
Add to Cart
Add to Cart
This item is not returnable. View return policy
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Sabouraud Dextrose Agar (Emmons) w/Chloramphenicol.
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Thermo Scientific™ Remel™ Sabouraud Dextrose Agar (Emmons) w/Chloramphenicol. Sabouraud Dextrose Agar (Emmons) is a modification of the original medium developed by Sabouraud in 18921. Emmons neutralized the pH, increasing it to 7.0, and reduced the concentration of dextrose from 40 to 20 g/l2. Chloramphenicol is added to the medium to make it inhibitory to bacterial overgrowth while allowing isolation of pathogenic fungi3.
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Sabouraud Dextrose Agar (Emmons) w/Chloramphenicol.
  • Ready-to-use – Convenience of a prepared media.
  • Suitable for isolation of pathogenic fungi from a material containing a large number of other fungi or bacteria.
  • Selective – Chloramphenicol, which is a broad-spectrum antibiotic, inhibits the growth of a range of gram-positive and gram-negative bacteria.
This medium contains casein, meat peptones, nitrogenous compounds and amino acids necessary for the growth of fungi. Dextrose is an energy source which serves to support the growth of fungi. Chloramphenicol is a broad-spectrum antibiotic which inhibit a range of Gram-positive and Gram-negative bacteria. Yeast extract may be added to the medium as a growth supplement to ensure the growth of fastidious fungi.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Sabouraud Dextrose Agar, Emmons w/Chloramphenicol
Format Monoplate Deep Pour
Product Type Agar
Quantity 10/Pk.