Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sabouraud Dextrose Broth, 2%

Catalog No. R064410 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 100 mL
100/Pk.
20/Pk.
Format:
15mm x 103mm Tube
WMB Lug - 100mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R064410 100/Pk. 15mm x 103mm Tube
R064412 20/Pk. 15mm x 103mm Tube
R112554 10 x 100 mL WMB Lug - 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R064410 Supplier Thermo Scientific™ Supplier No. R064410
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cultivates pathogenic yeast and fungi from clinical specimens with Sabouraud Dextrose Broth 2%.

Cultivates pathogenic yeast and fungi from clinical specimens with Thermo Scientific™ Remel™ Sabouraud Dextrose Broth 2%. Sabouraud Dextrose Agar is a modification of the original medium developed by Sabouraud in 18921. Emmons neutralized the pH, increasing it to 7.0, and reduced the concentration of dextrose from 40 to 20g/L2. Sabouraud Dextrose Broth 2% is a modification of the original formulation, without agar.

Cultivate pathogenic yeast and fungi from clinical specimens with Sabouraud Dextrose Broth 2%.

  • Ready-to-use – Convenience of a prepared media.
  • Suitable for isolation of pathogenic fungi from a material containing a large number of other fungi or bacteria.
  • Useful as an aid in the identification of yeasts3.

This medium contains casein and meat peptones nitrogenous compounds and amino acids necessary for the growth of yeasts and fungi. Dextrose is an energy source which serves to support the growth of fungi. The low pH of the medium is favorable to the growth of fungi, especially dermatophytes, while also inhibiting contaminating bacteria in clinical specimens.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Conforms to USP.
Description Sabouraud Dextrose Broth 2%
Format 15mm x 103mm Tube
Product Type Broth
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.