Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sabouraud Dextrose Broth, 2%
GSA_VA

Catalog No. R064412 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 100 mL
100/Pk.
20/Pk.
Format:
15mm x 103mm Tube
WMB Lug - 100mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R064412 20/Pk. 15mm x 103mm Tube
R064410 100/Pk. 15mm x 103mm Tube
R112554 10 x 100 mL WMB Lug - 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R064412 Supplier Thermo Scientific™ Supplier No. R064412
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cultivates pathogenic yeast and fungi from clinical specimens with Sabouraud Dextrose Broth 2%.

Cultivates pathogenic yeast and fungi from clinical specimens with Thermo Scientific™ Remel™ Sabouraud Dextrose Broth 2%. Sabouraud Dextrose Agar is a modification of the original medium developed by Sabouraud in 18921. Emmons neutralized the pH, increasing it to 7.0, and reduced the concentration of dextrose from 40 to 20g/L2. Sabouraud Dextrose Broth 2% is a modification of the original formulation, without agar.

Cultivate pathogenic yeast and fungi from clinical specimens with Sabouraud Dextrose Broth 2%.

  • Ready-to-use – Convenience of a prepared media.
  • Suitable for isolation of pathogenic fungi from a material containi

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Conforms to USP.
Description Sabouraud Dextrose Broth 2%
Format 15mm x 103mm Tube
Product Type Broth
Quantity 20/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.