Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sabouraud Dextrose Broth, 4%

Catalog No. R064416 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R064416 20 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R064416 Supplier Thermo Scientific™ Supplier No. R064416
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cultivate pathogenic yeast and molds from clinical specimens with Sabouraud Dextrose Broth 4%.

Cultivate pathogenic yeast and molds from clinical specimens with Thermo Scientific™ Remel™ Sabouraud Dextrose Broth 4%. Sabouraud Dextrose Broth is a modification of the dextrose agar described by Sabouraud1,2.

Cultivate pathogenic yeast and molds from clinical specimens with Sabouraud Dextrose Broth 4%.

  • Ready-to-use – Convenience of a prepared media.
  • Suitable for isolation of pathogenic molds from a material containing a large amount of other molds or bacteria.
  • Low pH makes the medium favorable for the growth of fungi especially dermatophytes, inhibiting some contaminating bacteria.

This medium contains casein and meat peptones nitrogenous compounds and amino acids necessary for the growth of yeasts and fungi. Dextrose is an energy source which serves to support the growth of fungi. The low pH of the medium is favorable to the growth of fungi, especially dermatophytes, while also inhibiting contaminating bacteria in clinical specimens.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Sabouraud Dextrose Broth 4% (pH 5.6)
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 20 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.