Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Salmonella Shigella Agar (SS Agar)
GSA_VA

Catalog No. r01840
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
Format:
12mm x 85mm Monoplate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R01840 10/Pk. 12mm x 85mm Monoplate
R01842 100/Pk. 12mm x 85mm Monoplate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R01840 Supplier Thermo Scientific™ Supplier No. R01840
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Use for the selective and differential isolation of Salmonella and to a lesser degree, Shigella spp.

Thermo Scientific™ Remel Salmonella Shigella Agar (SS Agar) is used for the selective and differential isolation of Salmonella and to a lesser degree, Shigella spp.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: COMPF, USDA.
Description Salmonella Shigella (SS) Agar
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.

Part No. R119756 not intended for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.