Description
Grow and isolate obligate and facultative anaerobic organisms, such as the fastidious anaerobic bacteria Bacteroides, Prevotella and Porphyromonas, using Thermo Scientific™ Remel™ Schaedler Anaerobe Agar with Sheep Blood, Haemin and Vitamin K1
Schaedler Agar w/Blood, Hemin, Vitamin K contains cysteine hydrochloride and glucose, as reducing substances, with the advantage that cysteine inhibits the growth of Escherichia coli. Schaedler Agar w/Blood, Hemin, Vitamin K has been shown to be a suitable alternative to blood agar for the enumeration of clostridia and has been used for the examination of food, waste products and ditch water. Studies have shown the necessity for strict anaerobic conditions for the successful recovery of obligate anaerobes when using this medium without the addition of blood.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Schaedler Agar w/Blood, Hemin, Vitamin K |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |