Description
Thermo Scientific™ Remel Selective Rapid Urea is used for detection of Helicobacter pylori in gastric mucosal biopsy specimens.
Specifications
Specifications
| Description | Selective Rapid Urea |
| Detectable Analytes | H. pylori |
| Type | H. pylori Testing Kit |
| Packaging Type | 24 Vials/Pk. |
| Quantity | 24 x 0.5 mL |