Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Selenite Broth Media for Automation (MFA) Tubes

Catalog No. r064503
Change view
Click to view available options
Quantity:
100/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R064503 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R064503 Supplier Thermo Scientific™ Supplier No. R064503
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Maximize laboratory productivity when selectively enriching for Salmonella with media tubes designed for automated specimen processing.

Take full advantage of your automated specimen processing systems with Thermo Scientific™ Remel™ Selenite Broth Media for Automation (MFA) Tubes. Formulated with the same high quality media used in traditional glass tubes, these plastic tubes are specifically designed to work with automated specimen processors. Maximize laboratory productivity and reduce the risk of human error when selectively enriching for Salmonella spp. by introducing complete specimen automation.

Selectively enrich for Salmonella from food and clinical samples using Remel Selenite Broth Media for Automation (MFA) Tubes. Selenite Broth is recommended for the isolation of Salmonella in Pharmacopeia and food testing guidelines.

  • Convenient: skirted design enables automated loading into specimen processors
  • Consistent: the traditional broth formula you are familiar with
  • Complementary: part of an extensive portfolio for your isolation, identification and differentiation workflow

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Cap Color White
Description Selenite Broth Media for Automation (MFA) Tubes
Format 12 x 80mm Tube
Product Type Prepared Microbiology Tubes
Fill Volume 4 mL
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.