Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Selenite Broth w/Cystine

Catalog No. R064508 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 9 mL
100 x 9 mL
100 x 10 mL
Format:
15mm x 103mm Tube
16mm x 125mm Tube
Packaging Type:
100/Pk.
20/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Packaging Type
R064508 20 x 9 mL 15mm x 103mm Tube 20/Pk.
R064506 100 x 9 mL 15mm x 103mm Tube 100/Pk.
R07841 100 x 10 mL 16mm x 125mm Tube 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R064508 Supplier Thermo Scientific™ Supplier No. R064508
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate Salmonella spp. using selective enrichment broth Selenite Broth with Cystine.

Thermo Scientific™ Remel Selenite Broth w/Cystine is a selective enrichment medium for isolation of Salmonella.

Leifson formulated Selenite Broth for enrichment of Salmonella from sources such as feces, urine, and food. Selenite was found to inhibit coliforms and fecal streptococci, however after 12-18 hours of incubation7 the breakthrough of

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP.
Description Selenite Broth w/Cystine
Format 15mm x 103mm Tube
Product Type Broth
Packaging Type 20/Pk.
Quantity 20 x 9 mL

REF R07841, REF R112570, and REF R112571 not intended for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.