Learn More
Thermo Scientific™ Remel™ Settling Plate, Sterile, Sabouraud Dextrose Agar w/Lecithin, Polysorbate 80
Catalog No. R111883 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
13mm x 85mm Plate - Double + Bag
Catalog No. Format
R111883 13mm x 85mm Plate - Double + Bag
1 options
This item is not returnable. View return policy
Catalog No. R111883 Supplier Thermo Scientific™ Supplier No. R111883
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel Settling Plate Sterile Sabouraud Dextrose Agar w/Lecithin and Polysorbate 80 facilitates cultivation of yeast and fungi.
Thermo Scientific™ Remel Settling Plate Sterile Sabouraud Dextrose Agar w/Lecithin and Polysorbate 80 facilitates cultivation of yeast and fungi.
  • Double bagged and gamma-irradiated with a third sterile bag enclosed for transporting plates to the laboratory
  • Sterile 85mm plates are manufactured in a cleanroom (SAL=106)
  • 100mm and 150mm plates are manufactured in an environmentally controlled environment
TRUSTED_SUSTAINABILITY
Specifications
Description Settling Plate Sterile Sabouraud Dextrose Agar w/Lecithin w/Polysorbate 80
Format 13mm x 85mm Plate - Double + Bag
Product Type Agar Plate
Quantity 10/Pk.
Not intended for IVD use.