Learn More
Thermo Scientific™ Settling Plate, Sterile, Tryptic Soy Agar (Irradiated)
GSA_VA
Catalog No. R111870 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
20/Pk.
Format:
150mm Plate - Double + Bag
Monoplate - Double + Bag
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R111870 10/Pk. Monoplate - Double + Bag
R111874 20/Pk. 150mm Plate - Double + Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R111870 Supplier Thermo Scientific™ Supplier No. R111870
Add to Cart
Add to Cart
This item is not returnable. View return policy
Isolate and cultivate wide variety of organisms like bacteria, yeast and fungi using Settling Plate Sterile Tryptic Soy Agar (Irradiated).
Isolate and cultivate wide variety of organisms like bacteria, yeast and fungi using Thermo Scientific™ Remel™ Settling Plate Sterile Tryptic Soy Agar (Irradiated). It is listed in Standard Methods for the Examination of Water and Wastewater and the Compendium of Methods for the Microbiological Examination of Foods1,2. It is also recommended for use in Official Methods of Analysis of AOAC International, the International Dairy Federation (IDF), and the United States Department of Agriculture (USDA)3,4. Tryptic Soy Agar slants may be used to cultivate, store, and ship bacterial cultures.
  • Settling plates (single bag and double bag) are manufactured in a clean room environment
  • This provides the highest level of sterility assurance with no compromise of growth characteristics
  • Each lot is tested to ensure consistent, reproducible results
  • Plate size:  85 x 13mm
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Settling Plate Sterile Tryptic Soy Agar
Format Monoplate - Double + Bag
Product Type Agar Plate
Quantity 10/Pk.
Dimensions (Dia. x H) 85 x 13 mm
Not intended for IVD use.