Learn More
Thermo Scientific™ Remel™ Settling Plate Tryptic Soy Agar with Lecithin and Polysorbate 80
Shop All Thermo Scientific ProductsDescription
- Sterile plated culture media
- Unique 100mm plate
- Double bagged typek pouch
- Convenient packaging for transport
- Confidence of no contamination in cleanroom
- Media that grows microorganisms which are compromised to confirm cleanroom standards
Specifications
Specifications
| Format | 100 mm Plate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.