Learn More
Thermo Scientific™ Settling Plate Tryptic Soy Agar
Catalog No. R111879 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
100mm Plate, Double
85mm Plate, Double
Settling Monoplate, Double
Settling Monoplate, Single
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
R111879 100mm Plate, Double
R111570 Settling Monoplate, Single
R111670 Settling Monoplate, Double
R111890 85mm Plate, Double
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R111879 Supplier Thermo Scientific™ Supplier No. R111879
Add to Cart
Add to Cart
Isolate and cultivate wide variety of organisms like bacteria, yeast and fungi using Settling Plate Tryptic Soy Agar.
Isolate and cultivate wide variety of organisms such as bacteria, yeast and fungi using Thermo Scientific™ Remel™ Settling Plate Tryptic Soy Agar. This medium is listed in Standard Methods for the Examination of Water and Wastewater and the Compendium of Methods for the Microbiological Examination of Foods1,2. This medium is recommended for use in Official Methods of Analysis of AOAC International, the International Dairy Federation (IDF), and the United States Department of Agriculture (USDA)3,4. Tryptic Soy Agar slants may be used to cultivate, store, and ship bacterial cultures.
Tryptic Soy Agar is prepared following the standard formula for Soybean-Casein Digest Agar Medium in the United States of Pharmacopeia (USP)5. Tryptic Soy Agar many be supplemented with blood to facilitate the growth of more fastidious bacteria or antimicrobial agents to permit the selection of various microbial groups from mixed flora.
  • Recommended for use in Official Methods of Analysis of AOAC International, IDF, and USDA3,4
  • Ready to use – Convenience of prepared medium
This medium contains casein and soy peptones that provide nitrogen, amino acids, and peptides necessary for bacterial growth. Dextrose is a ready source of energy. Sodium chloride supplies essential electrolytes and maintains osmotic equilibrium.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Settling Plate Tryptic Soy Agar
Format 100mm Plate, Double
Product Type Agar Plate
Quantity 10/Pk.
Not intended for IVD use.