Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Simmons Citrate Agar
GSA_VA

Catalog No. R060496
Change view
Click to view available options
Quantity:
20 x 5 mL
100 x 5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R060496 20 x 5 mL
R060494 100 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R060496 Supplier Thermo Scientific™ Supplier No. R060496
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Differentiate gram-negative bacteria based on citrate utilization using Simmons Citrate Agar.

Differentiate gram-negative bacteria based on citrate utilization using Thermo Scientific™ Remel™ Simmons Citrate Agar. In 1923, Koser developed a liquid medium to differentiate Escherichia coli and Enterobacter aerogenes1. The medium contained an inorganic ammonium salt and sodium citrate that provided a single nitrogen source and a single carbon source respectively. In 1926, this formulation was modified by Simmons who added 1.5% agar and brom thymol blue indicator2.

Gram-negative bacteria that can utilize ammonium dihydrogen phosphate as a sole nitrogen source and sodium citrate as a sole carbon source will grow on Citrate Agar (Simmons). These organisms extract nitrogen from the ammonium salt by breaking it down

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, USDA.
Description Simmons Citrate Agar
Format 15mm x 103mm Tube - Slant
Product Type Agar
Quantity 20 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.