Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Staphaurex™ Latex Agglutination Test
GSA_VA

Catalog No. p-4529287
Change view
Click to view available options
Quantity:
120 Tests/Kit
400 Tests/Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R30859902 400 Tests/Kit
R30859901 120 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R30859902 Supplier Thermo Scientific™ Supplier No. R30859902
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rapidly identify and differentiate staphylococci which possess coagulase and/or protein A, particularly Staphylococcus aureus, from other staphylococci.

Rapidly identify and differentiate staphylococci which possess coagulase and/or protein A, particularly Staphylococcus aureus, using the Thermo Scientific™ Staphaurex™ Latex Agglutination Test.

Experience quality and reliability with this rapid and accurate latex test for the identification and differentiatiation of staphylococcus aureus. White latex particles are coated with human fibrinogen for detection of clumping factor and specific IgG for detection of protein A.

When mixed on a slide with a suspension of S. aureus, reaction of clumping factor with the fibrinogen and/or protein A with the IgG causes rapid, strong agglutination of the latex particles.

Proven Performance
  • Rely on tried-and-true tests with more than 50 years of combined proven performance
  • Maintain confidence with reliable results that have greater than 99% sensitivity and specificity (according to data available in the package inserts)
Workflow Efficiency
  • Clear-cut, robust reactions provide easy-to-read results
  • Improve productivity with compact kit configurations, color-coded reagents, and convenient workstations
  • Flexible use from selective and non-selective media
  • Control latex is not required
Economical Solution
  • Maximize spending efficiency with the option of purchasing separate kit components
  • Save time, money, and resources by turning to one company, one source, for a complete set of Staph identification solutions
Although Staphylococci are commonly found on the skin and in mucous membranes, they have been associated with many human and animal infections. S.aureus and other coagulase-positive Staphylococci have been identified as a cause of suppurative infections, food poisoning and toxic shock syndrome and have been isolated from nearly all anatomical sites.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Control Sets Positive
Description Staphaurex Latex Agglutination Test
CE Marker Yes
Detectable Analytes S. aureus Coagulase, Protein A
Type Staphylococcus aureus Test Kit
Includes Test latex, disposable reaction cards, disposable sampling and mixing sticks
Quantity 400 Tests/Kit
Target Organism Class Staphylococcus aureus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.