Learn More
Description
Selectively isolate beta-haemolytic group A streptococci from throat cultures and upper respiratory specimens with this selective, ready-to-use medium.
Selectively isolate beta-hemolytic group A streptococci from throat cultures and upper respiratory specimens with Thermo Scientific™ Remel™ Strep A Isolation Agar. Several investigators have demonstrated improved isolation rates using sheep blood agar incorporated with selective agents to inhibit the commensal microbial flora commonly encountered in the oropharynx. In 1977, Gunn et al. developed a selective medium consisting of sheep blood agar supplemented with sulfamethoxazole and trimethoprim for isolating groups A and B streptococci1.
Streptococcus pyogenes is an important human pathogen that causes a wide variety of acute infections, such as soft tissue infections and pharyngitis; severe life-threatening infections, such as streptococcal toxic shock syndrome; and devastating post-infectious sequelae, such as rheumatic fever and glomerulonephritis.
Use Strep A Isolation Agar for selective isolation of beta-hemolytic group A streptococci from throat cultures and upper respiratory specimens.
- Selective: a unique combination of selective ingredients used in the medium suppresses the commensal microbial flora.
- Ready-to-use: convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Description | Strep A Isolation Agar |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.