Learn More
Thermo Scientific™ Strep Selective II Agar
Catalog No. R01859 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01859 100/Pk.
R01858 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R01859 Supplier Thermo Scientific™ Supplier No. R01859
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Selectively isolate beta-hemolytic Group A Streptococcus from throat cultures and upper respiratory specimens using Strep Selective II Agar.
Selectively isolate β-hemolytic Group A Streptococcus from throat cultures and upper respiratory specimens using Thermo Scientific™ Remel™ Strep Selective II Agar (w/colistin, oxolinic acid). Pacifico at al. demonstrated the isolation of β-hemolyic streptococci is improved with the use of a selective medium which inhibits oropharyngeal commensal microbial flora1. In 1984, Pelts developed a selective medium containing colistin and oxolinic acid2. This medium was valuable for isolation of streptococci from mixed flora, resulting in increased isolation rates and early identification of isolates.
Colistin and oxolinic acid allow growth of streptococci while inhibiting staphylococci, coryneforms, and gram-negative microorganisms.
TRUSTED_SUSTAINABILITY
Specifications
Description Strep Selective II Agar w/colistin, oxolinic acid
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 100/Pk.