Learn More
Thermo Scientific™ Streptex™ Acid Extraction Kit
GSA_VA
Catalog No. R30951301 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 Tests/Kit
Catalog No. Quantity
R30951301 50 Tests/Kit
1 options
This item is not returnable. View return policy
Catalog No. R30951301 Supplier Thermo Scientific™ Supplier No. R30951301
Add to Cart
Add to Cart
This item is not returnable. View return policy
Rapidly extract and identify streptococcal Lancefield group antigens from cultured organisms using Streptex™ Acid Extraction Kit.
Rapidly extract and identify streptococcal Lancefield group antigens from cultured organisms using Thermo Scientific™ Remel™ Streptex™ Acid Extraction Kit in conjunction with the Streptex™ Kit (ZL50/R30950501 and ZL61/R30164701). Lancefield showed that group-specific streptococcal antigens can be extracted in soluble form and identified by precipitation reactions with homologous antisera1. The Streptex™ kit requires at least 10 minutes incubation at 37°C to extract the group antigen. Reagents supplied in this kit allow a more rapid extraction of group antigen.
Species of Streptococcus are classified based on their hemolytic properties (alpha and beta). Beta-hemolytic streptococci are further classified by Lancefield grouping. The majority of species of Streptococcus possess group-specific antigens which are usually carbohydrate structural components of the cell wall.
Group-specific antigens are extracted from streptococci by nitrous acid in a simple and short incubation step. The extract is then neutralised and antigens are detected and identified with suspensions of latex particles coated with group-specific antibodies.
  • Ready-to-use: Reagent kit
  • Rapid extraction: Within 1 minute.
  • Ease of identification: Positive result is indicated by aggregation of latex particles.
The acid extraction kit provides reagents for the extraction of Lancefield group A, B, C, F and G at room temperature (18 – 30° C). Extraction is complete within 1 minute. Group D antigen cannot be extracted with these reagents.
The latex suspensions for groups A, B, C, D, F and G are supplied in the Streptex™ Kit (ZL50/R30950501 and ZL61/R30164701) or as individual suspensions.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Streptex and Acid Extraction Kit
CE Marker Yes
Detectable Analytes beta-haemolytic Streptococci
Type Streptococcus Test Kit
Includes 7mL each of Extraction Reagent 1, 2, and 3, disposable tubes, disposable tube holder, mixing sticks, sample dispensers
Organism Group Lancefield A, B, C, D, F and G
Quantity 50 Tests/Kit