Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Streptex™ Rapid Latex Agglutination Test

Catalog No. r30950555
Change view
Click to view available options
Quantity:
50 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R30950555 50 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R30950555 Supplier Thermo Scientific™ Supplier No. R30950555
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Isolate, identify and differentiate streptococci Lancefield groups A, B, C, F, and G with acid extraction for fast qualitative detection using the Thermo Scientific™ Streptex™ Rapid Latex Agglutination Test.

Streptex™ Rapid is a rapid latex test system for use in the qualitative detection and identification of the Lancefield group of streptococci. Reagents are provided for groups A, B, C, F and G.

Use this rapid latex agglutination test which provides a complete solution for the isolation and differentiation of Lancefield groups A, B, C, F & G. View distinct, clear results with white latex agglutination on a black background. Individual latex grouping reagents are available as required.

With Streptex, group specific antigens are extracted from streptococci in a simple incubation step. Antigens are then identified using polystyrene latex particles which have been coated with group-specific antibodies. These latex particles agglutinate strongly in the presence of homologous antigen and remain in smooth suspension in the absence of homologous antigen.

For a more rapid acid extraction method, Streptex Acid Extraction Kit (Part No. R30951301) is available for use with Streptex. Acid extraction will rapidly extract the Group antigens without the need for incubation. However, Group D antigen is not extracted efficiently with acid extraction; therefore, an alternative method must be used to confirm suspected Group D streptococci.

Proven Performance
  • Rely on tried-and-true tests with more than 50 years of combined proven performance
  • Maintain confidence with reliable results that have greater than 99% sensitivity (according to data available in the package insert)
Workflow Efficiency
  • Get easy-to-read results with clear-cut, robust reactions
  • Improve productivity with compact kit configurations, color-coded reagents and convenient workstations
  • Enjoy flexibility with use from selective and non-selective media—no control latex required
Economical Solution
  • Maximize spending efficiency with the option of purchasing separate kit components
  • Save time, money and resources by turning to one company, one source, for a complete set of Staph identification solutions
Streptococci are facultatively anaerobic, catalase-negative, gram-positive cocci that occur as pairs or chains. They cause a wide variety of clinical diseases including pharyngitis, scarlet fever, otitis media, impetigo, meningitis, neonatal sepis, pneumonia, endocarditis, osteomyelitis and necrotizing fascitis. One of the most useful phenotypic characteristics of streptococci is the hemolytic reaction, generally classified as alpha, beta or gamma, according to the appearance of zones around colonies growing on Blood Agar. Beta-hemolytic streptococci are further characterized by Lancefield groups based on the antigenic differences in group-specific polysaccharides located in the bacterial cell wall.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Control Sets Positive
Description Streptex Rapid Latex Agglutination Test
CE Marker Yes
Detectable Analytes Streptococci (Group A, B, C, F, G)
Type Streptococcus Test Kit
Includes Test latex reagents, extraction reagents, polyvalent positive control, disposable tubes, disposable tube holder, disposable reaction cards, mixing sticks and sample dispensers
Material Polyvalent
Organism Group Lancefield A, B, C, F and G
Quantity 50 Tests/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.