Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Stuart Transport Medium w/o Charcoal

Catalog No. r064622
Change view
Click to view available options
Quantity:
20 x 9 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R064622 20 x 9 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R064622 Supplier Thermo Scientific™ Supplier No. R064622
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Stuart Transport Medium is used to transport and preserve microbiological specimens.

Perform transport and preservation of microbiological specimens using Thermo Scientific™ Remel™ Stuart Transport Medium. Stuart originally described Stuart Transport Medium1 and Stuart et al. successfully used this medium to transport specimens intended for recovery of Neisseria gonorrhoeae to the laboratory2. Moffett et al. applied Stuart formlation to preserve viability of fastidious organisms such as Neisseria spp. during transport from clinic to laboratory3. Cooper used this medium to transport swabs containing upper respiratory tract specimens and fecal specimens4.

Stuart Transport Medium is a non-nutritional medium that maintains the viability of organisms without significant multiplication. Transport media are designed to preserve the viability of microorganisms when immediate inoculation of growth media is not possible. Methylene blue is an oxidation-reduction indicator and the medium in the reduced state appears colorless and in the oxidized state blue in color.

  • Ready-to-use – Convenience of prepared media
  • Isolates – Recovery of Neisseria gonorrhoeae

This medium contains small amount of agar that provides a semisolid consistency which prevents oxidation and drying during transport. Addition of sodium thioglycollate produces a reduced environment to improve the recovery of anaerobes. The permeability of bacterial cells is maintained by calcium chloride. Sodium glycerophosphate maintains the pH of the medium.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Stuart Transport Medium
Format 15mm x 103mm Tube - butt
Product Type Microbiology Media
Quantity 20 x 9 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.