Description
Selectively isolate beta-hemolytic Group A Streptococci from throat cultures and upper respiratory specimens using SXT Blood Agar.
Selectively isolate beta-hemolytic group A streptococci from throat cultures and upper respiratory specimens using Thermo Scientific™ Remel™ SXT Blood Agar. Pacifico at al. demonstrated the isolation of β-hemolyic streptococci is improved with the use of a selective medium which inhibits oropharyngeal commensal microbial flora1. In 1977, Gunn et al. introduced a selective medium containing sulfamethoxazole and trimethoprim in a tryptic soy agar base supplemented with 5% sheep blood2. In 1979, Kurzynski et al. proved that streptococci were more frequently isolated on SXT sheep blood agar3.
Clinical assessment alone is unreliable in distinguishing streptococcal and non-streptococcol pharyngitis4. Serious medical complications may follow pharyngitis caused by group A streptococci, making bacterial examination of the throat an indispensable diagnostic tool.
Use of SXT sheep blood agar allows enhanced recovery of Lancefield groups A and B streptococci from throat cultures while inhibiting the growth of commensal microbial flora.
- Ready-to-use – The convenience of prepared media
- Selective – Use of sulfamethoxazole plus trimethoprim (SXT) supresses growth of streptococcal groups C, F, and G and Streptococci viridans, most Enterobacteriaceae, Neisseria, and some Pseudomonas
This medium contains casein and soy peptones that provide nutritious source of nitrogen, amino acids, and peptides. Sheep blood provides growth factors and allows detection of hemolytic reactions. Sodium chloride maintains osmotic equilibrium. SXT (sulfamethoxazole, trimethoprim) act as selective agents to supresse most commensal microbial flora found in the oropharynx, including streptococcal groups C, F, and G and Streptococci viridans, most Enterobacteriaceae, Neisseria, and some Pseudomonas spp.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | SXT Blood Agar w/ sulfamethoxazole, trimethoprim |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |