Description
Selectively isolate beta-hemolytic Group A Streptococci from throat cultures and upper respiratory specimens using SXT Blood Agar.
Selectively isolate beta-hemolytic group A streptococci from throat cultures and upper respiratory specimens using Thermo Scientific™ Remel™ SXT Blood Agar. Pacifico at al. demonstrated the isolation of β-hemolyic streptococci is improved with the use of a selective medium which inhibits oropharyngeal commensal microbial flora1. In 1977, Gunn et al. introduced a selective medium containing sulfamethoxazole and trimethoprim in a tryptic soy agar base supplemented with 5% sheep blood2. In 1979, Kurzynski et al. proved that streptococci were more frequently isolated on SXT sheep blood agar3.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | SXT Blood Agar w/ sulfamethoxazole, trimethoprim |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 100/Pk. |