Description
Thermo Scientific™ Remel TB Auramine O Stain Kit is a fluorescent stain for microscopic detection of AFB.
Includes: 2 bottles Auramine O; 2 bottles Decolorizer; 2 bottles Potassium Permanganate.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 6/Kit |
| Includes | 2 bottles Auramine O; 2 bottles Decolorizer; 2 bottles Potassium Permanganate |
| Volume (Metric) | 250 mL/Bottle |
| Product Type | Auramine Stain |