Learn More
Thermo Scientific™ Remel™ TB Auramine-Rhodamine T Stain
GSA_VA
Catalog No. R40090 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5/Pk.
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R40090 Each
R40190 5/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R40090 Supplier Thermo Scientific™ Supplier No. R40090
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel TB Auramine-Rhodamine T is a fluorescent stain for microscopic detection of AFB.
Thermo Scientific™ Remel TB Auramine-Rhodamine T is a fluorescent stain for microscopic detection of AFB.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Quantity Each