Learn More
Thermo Scientific™ TB Digestant
Catalog No. R21266 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
6 x 100 mL
Catalog No. Quantity
R21266 6 x 100 mL
1 options
This item is not returnable. View return policy
Catalog No. R21266 Supplier Thermo Scientific™ Supplier No. R21266
This item is not returnable. View return policy
Digest and decontaminate clinical specimens prior to inoculation of media for isolation of Mycobacterium species using TB Digestant.
Digest and decontaminate clinical specimens prior to inoculation of media for isolation of Mycobacterium species with Thermo Scientific™ Remel™ TB Digestant. Respiratory specimens such as sputum contain mucin, which traps microorganisms. Such specimens require liquefaction to release the mycobacterial cells prior to inoculation of media. The N-acetyl-L-cysteine (NAC) method is the widely used procedure for digestion and decontamination of specimens for recovery of Mycobacterium spp1. TB Digestant has also been used successfully in Petroff's sodium hydroxide digestion method2.
The majority of clinical specimens submitted to the microbiology laboratory for isolation of acid-fast bacilli are contaminated with more rapidly growing commensal microbial flora. TB Digestant decontaminates the specimen by reducing or eliminating commensal microbial flora and digests respiratory secretions, releasing mycobacterial cells for detection in stains and cultures.
  • Ready-to-use – Liquid reagent supplied in 250mL bottle
  • Easy-to-store – Store at room temperature until used
TB Digestant contains 4% sodium hydroxide (CAS 1310-73-2) in demineralized water (CAS 7732-18-5). Specimens should remain in contact with the decontaminating solution for no longer than 15 minutes. Overprocessing may result in reduced recovery rates of Mycobacterium spp2.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Sputagest 100 Mucolytic Agent
Quantity 6 x 100 mL
Form Liquid