Description
Thermo Scientific™ Remel TB Kinyoun AFB Stain Kit is for use in qualitative procedures to differentiate acid-fast from non-acid-fast bacteria and for the detection of Cryptosporidium spp. in clinical specimens.
Includes: 2 bottles Kinyoun; 2 bottles Decolorizer; 2 bottles Methylene Blue.
Specifications
Specifications
| Quantity | 6 bottles/kit |
| Includes | 2 bottles Kinyoun; 2 bottles Decolorizer; 2 bottles Methylene Blue |
| Volume (Metric) | 250 mL/Bottle |
| Product Type | AFB Stain |