Description
Thermo Scientific™ Remel TB Kinyoun Carbolfuchsin is a primary stain for AFB and Cryptosporidium modified acid-fast staining procedures.
Specifications
Specifications
| Quantity | 5/Pk. |
| Volume (Metric) | 5 x 250 mL |