Learn More
Thermo Scientific™ Remel™ TB Methylene Blue Counterstain
GSA_VA
Catalog No. R40110 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5/Pk.
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R40110 Each
R40210 5/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R40110 Supplier Thermo Scientific™ Supplier No. R40110
Add to Cart
Add to Cart
This item is not returnable. View return policy
A counterstain for AFB and Cryptosporidium modified acid-fast stain using TB Kinyoun or TB Ziehl-Neelsen stains.
Thermo Scientific™ Remel TB Methylene Blue is a counterstain for AFB and Cryptosporidium modified acid-fast stain using TB Kinyoun or TB Ziehl-Neelsen stains.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Quantity Each