Description
Thermo Scientific™ Remel TB Methylene Blue is a counterstain for AFB and Cryptosporidium modified acid-fast stain using TB Kinyoun or TB Ziehl-Neelsen stains.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 5/Pk. |
| Volume (Metric) | 5 x 250 mL |