Learn More
Thermo Scientific™ TCBS Agar
GSA_VA
Catalog No. R01865 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
20mm x 150mm Pour Tube
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
R01865 12mm x 85mm Monoplate
R09832 20mm x 150mm Pour Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R01865 Supplier Thermo Scientific™ Supplier No. R01865
Add to Cart
Add to Cart
This item is not returnable. View return policy
Selectively isolate Vibrio cholerae and other enteropathogenic Vibrio spp. from clinical and non-clinical specimens using TCBS Agar.
Selectively isolate Vibrio cholerae and other enteropathogenic Vibrio spp. from clinical and non-clinical specimens using Thermo Scientific™ Remel™ TCBS Agar (Thiosulfate, Citrate, Bile Salts, Sucrose). In 1963, Kobayashi1 et al. modified Thiosulfate-Citrate-Bile Salts-Sucrose Agar (TCBS) developed by Nakanishi2. It was developed for selective isolation of vibrios which cause cholera, diarrhea, and food poisoning. In 1982, West et al. reported that TCBS Agar could be used for recovery of certain new pathogens such as Vibrio fluvialis and Vibrio vulnificus3.
TCBS Agar is a selective agar for the isolation and cultivation of Vibrio cholerae and other Vibrio spp. Vibrio cholerae is a gram-negative bacterium that causes cholera outbreaks and epidemics. Other Vibrio species
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF.
Description Thiosulfate Citrate Bile Salts Sucrose (TCBS) Agar
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.