Learn More
Thermo Scientific™ Tergitol 7 Agar w/TTC
Catalog No. R01875 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
Catalog No. Format
R01875 12mm x 85mm Monoplate
1 options
This item is not returnable. View return policy
Catalog No. R01875 Supplier Thermo Scientific™ Supplier No. R01875
This item is not returnable. View return policy
Selectively isolate and differentiate coliform organisms with ready-to-use Tergitol 7 Agar w/TTC.
Selectively isolate and differentiate coliform organisms with Thermo Scientific™ Remel™ Tergitol 7 Agar w/TTC. The use of tergitol (sodium heptadecyl sulfate) as a selective agent was first proposed by Pollard in 19461. Tergitol 7 Agar was later developed by Chapman for selective isolation and increased recovery of the coliform organisms2,3. Later Tergitol 7 Agar was modified by addition of triphenyltetrazolium chloride (TTC) to facilitate early recognition of Escherichia coli and Enterobacter aerogenes within 6-10 hours of incubation.
Triphenyltetrazolium chloride (TTC) aids in early detection of Escherichia coli and Enterobacter aerogenes.
TRUSTED_SUSTAINABILITY
Specifications
Description Tergitol 7 Agar w/TTC
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.