Learn More
Description
Selectively isolate Salmonella from fecal specimens using Tetrathionate Broth enrichment medium.
Selectively isolate Salmonella from fecal specimens using Thermo Scientific™ Remel™ Tetrathionate Broth enrichment medium. This broth was originally described by Muller for cultivation of Salmonella spp1. He demonstrated coliforms were inhibited in the medium while certain enteric gram-negative bacilli, such as Salmonella spp. grew very well. Kauffmann reported an increase in the recovery rate of Salmonella in the Muller medium2,3.
Salmonella are a group of closely related rod-shaped, gram-negative bacteria. Salmonella typically live in animal and human intestines and are shed through feces. Humans become infected most frequently through contaminated water or fo
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: AOAC, BAM, COMPF, SMWW, USDA, USP. |
| Description | Tetrathionate Broth |
| Format | E - 10mL |
| Product Type | Broth |
| Quantity | 20 x 10 mL |
Requires addition of Iodine Solution, REF R21228, REF R114350, or REF R114351. REF R112634 and REF R112630 not intended for IVD use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.