Learn More
Thermo Scientific™ Thayer Martin Agar Improved
GSA_VA
Catalog No. R01884 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01884 10/Pk.
R01886 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R01884 Supplier Thermo Scientific™ Supplier No. R01884
Add to Cart
Add to Cart
This item is not returnable. View return policy
Isolate Neisseria gonorrhoeae and Neisseria meningitidis using Thayer Martin Agar Improved.
Isolate Neisseria gonorrhoeae and Neisseria meningitidis with Thermo Scientific™ Remel Thayer Martin Agar Improved. Johnston introduced a medium to grow N. gonorrhoeae in 24 hours1 and this was the advent of GC Agar Base. Thayer and Martin formulated a selective medium to isolate N. gonorrhoeae and N. meningitidis that contained antibiotics capable of supressing the commensal microbial flora present in the anal canal, vagina, pharynx and other sites2. Martin and Lewis recommended the use of anisomycin to replace nystatin and increase the shelf life of the media3.
Various studies have demonstrated improved recovery for pathogenic Neisseria spp. using selective media4-6. Thayer-Martin agar Improved is a selective medium used for the isolation of Neisseria gonorrheae and N. meningitidis from specimens containing a mixed flora of bacteria and/or fungi. Thayer Martin Agar Improved contains hemoglobin, which provides the X factor (hemin), and GCHI Enrichment, which provides the V factor, vitamins, amino acids, coenzymes, and dextrose. Initially the antibiotics ristocetin and polymyxin B were used and replaced with vancomycin, colistimethate, and nystatin7.
  • Ready to use – Convenience of prepared medium.
  • Selective media –
    • Anisomycin suppresses Candida albicans.
    • Colistin inhibits most gram-negative microorganisms, including Pseudomonas spp.
    • Vancomycin inhibits gram-positive cocci.
    • Trimethoprim lactate suppresses swarming of Proteus spp.
The tablet provided in the JEMBEC™ and Pill Pocket systems, when coming into contact with the water vapor present in the medium, liberates carbon dioxide into the enclosed pack.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Thayer Martin Agar Improved
Format 12mm x 85mm Monoplate
Product Type Agar
Quantity 10/Pk.