Learn More
Thermo Scientific™ Thayer Martin Agar Improved
Description
Isolate Neisseria gonorrhoeae and Neisseria meningitidis using Thayer Martin Agar Improved.
Isolate Neisseria gonorrhoeae and Neisseria meningitidis with Thermo Scientific™ Remel Thayer Martin Agar Improved. Johnston introduced a medium to grow N. gonorrhoeae in 24 hours1 and this was the advent of GC Agar Base. Thayer and Martin formulated a selective medium to isolate N. gonorrhoeae and N. meningitidis that contained antibiotics capable of supressing the commensal microbial flora present in the anal canal, vagina, pharynx and other sites2. Martin and Lewis recommended the use of anisomycin to replace nystatin and increase the shelf life of the media3.
Various studies have demonstrated improved recovery for pathogenic Neisseria spp. using selective media4-6. Thayer-Martin agar Improved is a selective medium used for the isolation of Neisseria gonorrheae and N. meningitidis from specimens containing a mixed flora of bacteria and/or fungi. Thayer Martin Agar Improved contains hemoglobin, which provides the X factor (hemin), and GCHI Enrichment, which provides the V factor, vitamins, amino acids, coenzymes, and dextrose. Initially the antibiotics ristocetin and polymyxin B were used and replaced with vancomycin, colistimethate, and nystatin7.
- Ready to use – Convenience of prepared medium.
- Selective media –
- Anisomycin suppresses Candida albicans.
- Colistin inhibits most gram-negative microorganisms, including Pseudomonas spp.
- Vancomycin inhibits gram-positive cocci.
- Trimethoprim lactate suppresses swarming of Proteus spp.
The tablet provided in the JEMBEC™ and Pill Pocket systems, when coming into contact with the water vapor present in the medium, liberates carbon dioxide into the enclosed pack.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Description | Thayer Martin Agar Improved |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar |
| Quantity | 10/Pk. |