Learn More
Thermo Scientific™ Thayer Martin Agar Modified (w/nystatin, colistin, vancomycin, trimethoprim)
GSA_VA
Catalog No. R01882 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01882 100/Pk.
R01880 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R01882 Supplier Thermo Scientific™ Supplier No. R01882
Add to Cart
Add to Cart
This item is not returnable. View return policy
Isolate and identify Neisseria gonorrhoeae and Neisseria meningitidis with this selective, ready-to-use solid medium
Isolate Neisseria gonorrhoeae and Neisseria meningitidis with Thermo Scientific™ Remel™ Thayer Martin Agar Modified (with nystatin, colistin, vancomycin, trimethoprim)
Thayer Martin Agar is a selective medium containing antibiotics to isolate Neisseria gonorrhoeae and Neisseria meningitidis, while suppressing commensal microbial flora present in the anal canal, vagina, pharynx and other sites1
  • Nystatin inhibits Candida albicans
  • Colistin inhibits most gram-negative microorganisms, including Pseudomonas spp.
  • Vancomycin inhibits gram-positive cocci
  • Trimethoprim suppresses swarming of Proteus spp.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Thayer Martin Agar Modified
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 100/Pk.