Learn More
Thermo Scientific™ Tinsdale Enrichment
Catalog No. R45045 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 3.3 mL
Catalog No. Quantity
R45045 10 x 3.3 mL
1 options
This item is not returnable. View return policy
Catalog No. R45045 Supplier Thermo Scientific™ Supplier No. R45045
This item is not returnable. View return policy
Differentiate Corynebacterium species with Thermo Scientific™ Remel™ Tinsdale Enrichment when used with Tinsdale Agar Base (R09897).
Differentiate Corynebacterium species with Thermo Scientific™ Remel™ Tinsdale Enrichment when used with Tinsdale Agar Base (R09897).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Tinsdale Enrichment
Form Liquid
Product Type Enrichment
Quantity 10 x 3.3 mL