Learn More
Thermo Scientific™ Remel™ Todd Hewitt Broth w/Gentamicin, Nalidixic Acid, Blood
Shop All Thermo Scientific ProductsDescription
Todd Hewitt Broth with Gentamicin, Nalidixic Acid and Blood (Trans-Vag Broth w/5% Sheep Blood) is used for selective isolation of group B streptococci from clinical specimens containing mixed flora.
- Gentamicin and nalidixic acid inhibit some gram-negative bacteria
Specifications
Specifications
| Description | Todd Hewitt Broth; Trans-Vag Broth w/Blood; |
| Format | 15mm x 103mm Tube |
| Product Type | Microbiology Media |
| Quantity | 100 x 5 mL |