Learn More
Thermo Scientific™ Remel™ Trichrome Stain Kit
GSA_VA
Catalog No. R40217 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
R40217 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. R40217 Supplier Thermo Scientific™ Supplier No. R40217
This item is not returnable. View return policy
Thermo Scientific™ Remel Trichrome Stain Kit is used for trichrome staining.
Thermo Scientific™ Remel Trichrome Stain Kit is used for trichrome staining.
Includes:
250mL Trichrome Stain (Wheatley); 3 x 250mL Ethanol 70%; 250mL Acid Ethanol 90%; 2 x 250mL Ethanol 95%; 2 x 250mL Xylene-S; 5 x 0.75mL Lugol's Iodine ampules.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Includes 250mL Trichrome Stain (Wheatley); 3 x 250mL Ethanol 70%; 250mL Acid Ethanol 90%; 2 x 250mL Ethanol 95%; 2 x 250mL Xylene-S; 5 x 0.75mL Lugol's Iodine ampules.
Product Type Trichome Stain Kit