Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Triple Sugar Iron Agar
GSA_VA

Catalog No. R064850 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
Format:
15mm x 103mm Tube - Slant
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R064850 100/Pk. 15mm x 103mm Tube - Slant
R064852 20/Pk. 15mm x 103mm Tube - Slant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R064850 Supplier Thermo Scientific™ Supplier No. R064850
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides differentiation of gram-negative enteric bacilli based on carbohydrate fermentation and hydrogen sulfide production.

Thermo Scientific™ Remel Triple Sugar Iron Agar (TSI) provides differentiation of gram-negative enteric bacilli based on carbohydrate fermentation and hydrogen sulfide production.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, USDA, USP.
Description Triple Sugar Iron Agar
Format 15mm x 103mm Tube - Slant
Product Type Agar Slants
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.