Learn More
Thermo Scientific™ Remel™ Triton X-100 Mucolytic Reagent
Catalog No. R21902 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
15 mL
Catalog No. Quantity
R21902 15 mL
1 options
This item is not returnable. View return policy
Catalog No. R21902 Supplier Thermo Scientific™ Supplier No. R21902
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel Triton X-100 is a mucolytic reagent for fecal concentration procedures.
Thermo Scientific™ Remel Triton X-100 is a mucolytic reagent for fecal concentration procedures.
  • Polypropylene dispenser bottle
TRUSTED_SUSTAINABILITY
Specifications
Description Triton X-100
Quantity 15 mL
Form Liquid