Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Tryptic Soy Agar
GSA_VA

Catalog No. R064860 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 100 mL
10 x 500 mL
10/Pk.
100 x 22 mL
100/Pk.
20 x 22 mL
20/Pk.
Format:
15mm x 103mm Tube
15mm x 150mm Plate
20mm x 113mm Tube
20mm x 150mm Pour Tube
BB - 500mL
BRB - 100mL
Monoplate
Monoplate Deep Pour
Packaging Type:
10/Pk.
100/Pk.
20/Pk.
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Packaging Type
R064860 100/Pk. 15mm x 103mm Tube 100/Pk.
R01917 100/Pk. Monoplate 100/Pk.
R01920 10/Pk. Monoplate 10/Pk.
R04080 10/Pk. 15mm x 150mm Plate 10/Pk.
R064862 20/Pk. 15mm x 103mm Tube 20/Pk.
R09938 100 x 22 mL 20mm x 150mm Pour Tube 100/Pk.
R112700 10 x 500 mL BB - 500mL 10/Pk.
R112703 10 x 100 mL BRB - 100mL 10/Pk.
R01922 10/Pk. Monoplate Deep Pour 10/Pk.
R08930 100/Pk. 20mm x 113mm Tube 100/Pk.
R08932 20/Pk. 20mm x 113mm Tube 20/Pk.
R09940 20 x 22 mL 20mm x 150mm Pour Tube 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Catalog No. R064860 Supplier Thermo Scientific™ Supplier No. R064860
Add to Cart
Edge
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'

The Thermo Scientific™ Remel Tryptic Soy Agar (TSA) (Soybean Casein Digest Agar) is a general purpose medium for cultivation and isolation of microorganisms.

Thermo Scientific™ Remel Tryptic Soy Agar (TSA) (Soybean Casein Digest Agar) is a general purpose medium for cultivation and isolation of microorganisms.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
Description Tryptic Soy Agar
Format 15mm x 103mm Tube
Product Type Agar
Packaging Type 100/Pk.
Quantity 100/Pk.

REF R110092, REF R112700, and REF R112703 not intended for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.