Learn More
Thermo Scientific™ Remel™ Tryptic Soy Agar
Description
Thermo Scientific™ Remel Tryptic Soy Agar (TSA) (Soybean Casein Digest Agar) is a general purpose medium for cultivation and isolation of microorganisms.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: AOAC, BAM, COMPF, IDF, USDA, SMWW. |
| Description | Tryptic Soy Agar |
| Format | Monoplate |
| Product Type | Agar |
| Packaging Type | 100/Pk. |
| Quantity | 100/Pk. |
REF R110092, REF R112700, and REF R112703 not intended for IVD use.